accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CClClN

CAS RN 53342-53-3

IUPAC Name: 4,6-dichloro-2-methylquinoline
Synonyms: 4,6-dichloro-2-methylquinoline
Molecular Weight (g/mol): 212.07
Molecular Formula: C10H7Cl2N
InChi Key: KYBCQZHRJZJYCE-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C2C=C(Cl)C=CC2=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results