accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3

CAS RN 5337-93-9

IUPAC Name: 1-(4-methylphenyl)propan-1-one
Synonyms: 4'-methylpropiophenone
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: PATYHUUYADUHQS-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(C)C=C1
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)