Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Methylpropiophenone, 94%

Catalog No. AAB2167336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2167336 500 g
AAB2167322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2167336 Supplier Thermo Scientific Chemicals Supplier No. B2167336
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 5337-93-9 | C10H12O | 148.21 g/mol

4?-Methylpropiophenone is a chemical reagent used in electrocarboxylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methylpropiophenone is a chemical reagent used in electrocarboxylation reactions.

Solubility
Soluble in chloroform and hexane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5337-93-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00009312
InChI Key PATYHUUYADUHQS-UHFFFAOYSA-N
Synonym 4'-methylpropiophenone, 1-p-tolyl propan-1-one, p-methylpropiophenone, 4-methylpropiophenone, 1-4-methylphenyl propan-1-one, 1-propanone, 1-4-methylphenyl, p-tolyl ethyl ketone, p-methyl propiophenone, 1-p-tolylpropan-1-one, 1-4-methylphenyl-1-propanone
PubChem CID 21429
ChEBI CHEBI:59335
IUPAC Name 1-(4-methylphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 7°C
Density 0.991
Boiling Point 238°C to 239°C
Flash Point 108°C (226°F)
Refractive Index 1.528
Quantity 500 g
Beilstein 2042137
Solubility Information Soluble in chloroform and hexane.
Formula Weight 148.21
Percent Purity 94%
Chemical Name or Material 4'-Methylpropiophenone
Show More Show Less
EINECSNumber 226-267-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.