accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3(E)

CAS RN 53398-80-4

IUPAC Name: (2E)-hex-2-en-1-yl propanoate
Synonyms: trans-2-hexenyl propionate
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: LPWKTEHEFDVAQS-VOTSOKGWSA-N
SMILES: CCC\C=C\COC(=O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results