Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hexen-1-yl Propionate 95.0+%, TCI America™
SDP

Catalog No. P088425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P088425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P088425ML Supplier TCI America Supplier No. P088425ML

Chemical Identifiers

CAS 53398-80-4
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00036545
InChI Key LPWKTEHEFDVAQS-VOTSOKGWSA-N
Synonym Propionic Acid trans-2-Hexen-1-yl Ester
PubChem CID 5352463
ChEBI CHEBI:87682
IUPAC Name [(E)-hex-2-enyl] propanoate
SMILES CCCC=CCOC(=O)CC

Specifications

Color Colorless
Boiling Point 90°C
Quantity 25 mL
Formula Weight 156.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hexen-1-yl Propionate
EINECSNumber (2)-0760
RTECSNumber MP8645000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.