accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53398-83-7

H3COOCH3(E)

CAS RN 53398-83-7

IUPAC Name: (2E)-hex-2-en-1-yl butanoate
Synonyms: trans-2-hexenyl butyrate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: PCGACKLJNBBQGM-VOTSOKGWSA-N
SMILES: CCC\C=C\COC(=O)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results