accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 534-15-6

IUPAC Name: 1,1-dimethoxyethane
Synonyms: 1,1-dimethoxyethane
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: SPEUIVXLLWOEMJ-UHFFFAOYSA-N
SMILES: COC(C)OC
Molecular Weight (g/mol) 
  • (3)
  • (6)
Boiling Point 
  • (3)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)