accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaSSS

CAS RN 534-18-9

IUPAC Name: disodium (sulfanidylmethanethioyl)sulfanide
Synonyms: disodium trithiocarbonate
Molecular Weight (g/mol): 154.17
Molecular Formula: CNa2S3
InChi Key: USAIOOFEIMNEDN-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[S-]C([S-])=S
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)