Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiocarbonate, 40% in Water, Thermo Scientific™

Catalog No. AC419661000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419661000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC419661000 Supplier Thermo Scientific Chemicals Supplier No. 419661000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-18-9
Molecular Formula CNa2S3
Molecular Weight (g/mol) 154.171
InChI Key USAIOOFEIMNEDN-UHFFFAOYSA-L
Synonym sodium trithiocarbonate, carbonotrithioic acid, disodium salt, disodium carbonotrithioate, unii-y01pt1as4o, disodium trithiocarbonate, y01pt1as4o, carbonotrithioic acid, sodium salt 1:2, sodium carbonotrithioate, d0d5xc, trithiocarbonic acid disodium salt
PubChem CID 2735136
IUPAC Name disodium;carbonotrithioate
SMILES C(=S)([S-])[S-].[Na+].[Na+]

Specifications

Color Orange-Red
Linear Formula Na2CS3
Merck Index 15, 8825
Formula Weight 154.19
Percent Purity 37 to 43% (NA2CS3)
Quantity 100 g
Physical Form Solution
Chemical Name or Material Sodium thiocarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.