accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CNHHN

CAS RN 534-26-9

IUPAC Name: 2-methylimidazolidin-2-ylium
Synonyms: lysidine (chemical)
Molecular Weight (g/mol): 85.13
Molecular Formula: C4H9N2
InChi Key: VWSLLSXLURJCDF-UHFFFAOYSA-O
SMILES: C[C+]1NCCN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results