Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-imidazoline 97.0+%, TCI America™
SDP

Catalog No. M057925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M057925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M057925G Supplier TCI America Supplier No. M057925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 534-26-9
Molecular Formula C4H8N2
Molecular Weight (g/mol) 84.122
MDL Number MFCD00051490
InChI Key VWSLLSXLURJCDF-UHFFFAOYSA-N
Synonym 2-methyl-2-imidazoline, 2-methylimidazoline, 1h-imidazole, 4,5-dihydro-2-methyl, methylglyoxalidine, 2-imidazoline, 2-methyl, 4,5-dihydro-2-methyl-1h-imidazole, unii-987f50e3pe, 2-methyl imidazoline, 2-methyl-4,5-dihydroimidazole, 2-methyl-20imidazolin
PubChem CID 10798
IUPAC Name 2-methyl-4,5-dihydro-1H-imidazole
SMILES CC1=NCCN1

Specifications

Melting Point 104°C
Color White-Yellow
Boiling Point 130°C
Quantity 25 g
Formula Weight 84.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-2-imidazoline
EINECSNumber (5)-0422
RTECSNumber NI4804995
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.