CAS RN 5341-61-7
Hydrazine dihydrochloride, 99%
CAS: 5341-61-7 Molecular Formula: ClH5N2 Molecular Weight (g/mol): 68.50 MDL Number: MFCD00064543 InChI Key: BIVUUOPIAYRCAP-UHFFFAOYSA-N Synonym: hydrazine dihydrochloride,hydrazine, dihydrochloride,unii-km8dd9j0c8,ccris 5479,diamine dihydrochloride,hydrazinium monochloride,km8dd9j0c8,hydrazine, hydrochloride 1:2,hydrazinedihydrochloride,hydrazine hcl PubChem CID: 17548 IUPAC Name: hydrazine;dihydrochloride SMILES: [H+].[Cl-].NN
Hydrazine Standard, 1000 ppm N2H4, Ricca Chemical
CAS: 5341-61-7 Molecular Formula: ClH5N2 Molecular Weight (g/mol): 68.50 MDL Number: MFCD00064543 InChI Key: BIVUUOPIAYRCAP-UHFFFAOYSA-N PubChem CID: 17548 IUPAC Name: hydrogen hydrazine chloride SMILES: [H+].[Cl-].NN
Hydrazine dihydrochloride, 98+%
CAS: 5341-61-7 Molecular Formula: ClH5N2 Molecular Weight (g/mol): 68.50 MDL Number: MFCD00064543 InChI Key: BIVUUOPIAYRCAP-UHFFFAOYSA-N Synonym: hydrazine dihydrochloride,hydrazine, dihydrochloride,unii-km8dd9j0c8,ccris 5479,diamine dihydrochloride,hydrazinium monochloride,km8dd9j0c8,hydrazine, hydrochloride 1:2,hydrazinedihydrochloride,hydrazine hcl PubChem CID: 17548 IUPAC Name: hydrazine;dihydrochloride SMILES: [H+].[Cl-].NN
Hydrazine Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N2H4), LabChem™
CAS: 5341-61-7 Molecular Formula: ClH5N2 Molecular Weight (g/mol): 68.50 MDL Number: MFCD00064543 InChI Key: BIVUUOPIAYRCAP-UHFFFAOYSA-N PubChem CID: 17548 IUPAC Name: hydrogen hydrazine chloride SMILES: [H+].[Cl-].NN
Hydrazine Standard, 100 ppm N2H4, Ricca Chemical
CAS: 5341-61-7 Molecular Formula: ClH5N2 Molecular Weight (g/mol): 68.50 MDL Number: MFCD00064543 InChI Key: BIVUUOPIAYRCAP-UHFFFAOYSA-N PubChem CID: 17548 IUPAC Name: hydrogen hydrazine chloride SMILES: [H+].[Cl-].NN
Hydrazine Dihydrochloride 98.0+%, TCI America™
CAS: 5341-61-7 Molecular Formula: ClH5N2 Molecular Weight (g/mol): 68.50 MDL Number: MFCD00064543 InChI Key: BIVUUOPIAYRCAP-UHFFFAOYSA-N Synonym: hydrazine dihydrochloride,hydrazine, dihydrochloride,unii-km8dd9j0c8,ccris 5479,diamine dihydrochloride,hydrazinium monochloride,km8dd9j0c8,hydrazine, hydrochloride 1:2,hydrazinedihydrochloride,hydrazine hcl PubChem CID: 17548 IUPAC Name: hydrogen hydrazine chloride SMILES: [H+].[Cl-].NN