Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Dihydrochloride (Crystalline/Certified), Fisher Chemical

Catalog No. H319500 Shop All Fisherbrand Products
Change view
Click to view available options
Poly Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
H319-500 Poly Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H319-500

missing translation for 'validMainframeMsg'

Specifications

Boiling Point 200°C
Melting Point 198°C
Color White
CAS 5341-61-7
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range ≥98 %
Molecular Formula ClH5N2
Linear Formula NH2NH2·2HCl
MDL Number MFCD00064543
Synonym hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
SMILES [H+].[Cl-].NN
IUPAC Name hydrogen hydrazine chloride
Molecular Weight (g/mol) 68.50
PubChem CID 17548
Formula Weight 104.97
Percent Purity ≥98%
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Hydrazine Dihydrochloride
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.