accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)O(S)HO(R)HO

CAS RN 53448-53-6

IUPAC Name: (2R,3R,4S)-2-(hydroxymethyl)oxolane-3,4-diol
Synonyms: anhydroxylitol
Molecular Weight (g/mol): 134.13
Molecular Formula: C5H10O4
InChi Key: KZVAAIRBJJYZOW-VPENINKCSA-N
SMILES: OC[C@H]1OC[C@H](O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results