Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Anhydro-D-xylitol 97.0+%, TCI America™
SDP

Catalog No. A263520MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A263520MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A263520MG Supplier TCI America Supplier No. A263520MG

Specifications

CAS 53448-53-6
Color Yellow
Boiling Point 160°C
Molecular Formula C5H10O4
MDL Number MFCD27923011
Quantity 20 mg
InChI Key KZVAAIRBJJYZOW-VPENINKCSA-N
SMILES OC[C@H]1OC[C@H](O)[C@H]1O
IUPAC Name (2R,3R,4S)-2-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol) 134.13
PubChem CID 104501
Formula Weight 134.13
Percent Purity ≥97.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1,4-Anhydro-D-xylitol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.