accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOClHNNO

CAS RN 53449-14-2

IUPAC Name: 7-chloro-6-nitro-1,4-dihydroquinazolin-4-one
Synonyms: 7-chloro-6-nitro-1H-quinazolin-4-one
Molecular Weight (g/mol): 225.59
Molecular Formula: C8H4ClN3O3
InChi Key: URDYTQYZXZKBQT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C=C2NC=NC(=O)C2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)