accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5351-69-9

IUPAC Name: 3-amino-1-phenylthiourea
Synonyms: 4-phenylthiosemicarbazide
Molecular Weight (g/mol): 167.23
Molecular Formula: C7H9N3S
InChi Key: KKIGUVBJOHCXSP-UHFFFAOYSA-N
SMILES: NNC(=S)NC1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)