Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-3-thiosemicarbazide, 99%, Thermo Scientific™

Catalog No. AC259741000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC259741000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC259741000 Supplier Thermo Scientific Chemicals Supplier No. 259741000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5351-69-9
Molecular Formula C7H9N3S
Molecular Weight (g/mol) 167.23
MDL Number MFCD00007615
InChI Key KKIGUVBJOHCXSP-UHFFFAOYSA-N
Synonym 4-phenyl-3-thiosemicarbazide, 4-phenylthiosemicarbazide, n-phenylhydrazinecarbothioamide, hydrazinecarbothioamide, n-phenyl, phenyl thiosemicarbazide, semicarbazide, 4-phenyl-3-thio, 4-phenyl thiosemicarbazide, usaf el-45, usaf ek-5426, 3-amino-1-phenylthiourea
PubChem CID 730679
IUPAC Name 1-amino-3-phenylthiourea
SMILES C1=CC=C(C=C1)NC(=S)NN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 138°C to 141°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H5NHCSNHNH2
Quantity 100 g
Beilstein 12, 412
Formula Weight 167.24
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Phenyl-3-thiosemicarbazide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.