accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3COOFFF

CAS RN 53518-14-2

IUPAC Name: 7-(dimethylamino)-4-(trifluoromethyl)-2H-chromen-2-one
Synonyms: coumarin 152
Molecular Weight (g/mol): 257.21
Molecular Formula: C12H10F3NO2
InChi Key: KDTAEYOYAZPLIC-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results