Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(Dimethylamino)-4-(trifluoromethyl)coumarin 97.0+%, TCI America™
SDP

Catalog No. D33565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33565G Supplier TCI America Supplier No. D33565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53518-14-2
Molecular Formula C12H10F3NO2
Molecular Weight (g/mol) 257.21
MDL Number MFCD00041867
InChI Key KDTAEYOYAZPLIC-UHFFFAOYSA-N
Synonym coumarin 152, 7-dimethylamino-4-trifluoromethyl coumarin, 2h-1-benzopyran-2-one, 7-dimethylamino-4-trifluoromethyl, 7-dimethylamino-4-trifluoromethylcoumarin, 7-dimethylamino-4-trifluoromethyl chromen-2-one, 7-dimethylamino-4-trifluoromethyl-2h-chromen-2-one, coumarin-152, acmc-209l8g, 7-nme2-4-cf3-coumarin, 7-dimethylamino-4-trifluoromethyl-2-benzopyrone
PubChem CID 104522
ChEBI CHEBI:51775
IUPAC Name 7-(dimethylamino)-4-(trifluoromethyl)-2H-chromen-2-one
SMILES CN(C)C1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1

Specifications

Melting Point 147°C
Quantity 5 g
Formula Weight 257.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Dimethylamino)-4-(trifluoromethyl)coumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.