accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHNNHO

CAS RN 5354-94-9

IUPAC Name: 3-(1H-imidazol-5-yl)-2-(phenylformamido)propanoic acid
Synonyms: 3-(3H-imidazol-4-yl)-2-(phenylformamido)propanoic acid
Molecular Weight (g/mol): 259.27
Molecular Formula: C13H13N3O3
InChi Key: AUDPUFBIVWMAED-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CN=CN1)NC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results