Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl-L-histidine Monohydrate 98.0+%, TCI America™
SDP

Catalog No. B02041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02041G 1 g
B0204100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B02041G Supplier TCI America Supplier No. B02041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5354-94-9
Molecular Formula C13H13N3O3
Molecular Weight (g/mol) 259.265
MDL Number MFCD00037850
InChI Key AUDPUFBIVWMAED-NSHDSACASA-N
PubChem CID 152263
IUPAC Name (2S)-2-benzamido-3-(1H-imidazol-5-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CC2=CN=CN2)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 259.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyl-L-histidine Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.