accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5355-68-0

IUPAC Name: 4-oxo-1-(propan-2-yl)piperidin-1-ium
Synonyms: 1-isopropyl-4-oxopiperidin-1-ium
Molecular Weight (g/mol): 142.22
Molecular Formula: C8H16NO
InChi Key: CCDBCHAQIXKJCG-UHFFFAOYSA-O
SMILES: CC(C)[NH+]1CCC(=O)CC1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)