Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropyl-4-piperidone 97.0+%, TCI America™
SDP

Catalog No. I05665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05665G 5 g
I056625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05665G Supplier TCI America Supplier No. I05665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5355-68-0
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00038035
InChI Key CCDBCHAQIXKJCG-UHFFFAOYSA-N
Synonym 1-isopropyl-4-piperidone, 1-isopropylpiperidin-4-one, n-isopropyl-4-piperidone, 4-piperidinone, 1-1-methylethyl, 1-isopropyl-4-piperidinone, 4-piperidone, 1-isopropyl, 1-propan-2-yl piperidin-4-one, 1-1'-methylethyl-4-piperidone, 1-methylethyl piperidin-4-one
PubChem CID 79313
IUPAC Name 1-propan-2-ylpiperidin-4-one
SMILES CC(C)N1CCC(=O)CC1

Specifications

Boiling Point 101°C
Quantity 5 g
Formula Weight 141.21
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Isopropyl-4-piperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.