accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3COONH3CH3CNHOHCl

CAS RN 53567-47-8

IUPAC Name: 3-{4-[2-(diethylamino)ethoxy]phenyl}-2-(phenylformamido)-N,N-dipropylpropanamide hydrochloride
Synonyms: tiropramide hydrochloride
Molecular Weight (g/mol): 504.11
Molecular Formula: C28H42ClN3O3
InChi Key: SVCQSEFEZWMYSA-UHFFFAOYNA-N
SMILES: Cl.CCCN(CCC)C(=O)C(CC1=CC=C(OCCN(CC)CC)C=C1)NC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results