accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53597-69-6

IUPAC Name: 1,3,5-triaza-7-phosphatricyclo[3.3.1.1³,⁷]decane
Synonyms: 1,3,5-triaza-7-phosphatricyclo[3.3.1.1³,⁷]decane
Molecular Weight (g/mol): 157.16
Molecular Formula: C6H12N3P
InChi Key: FXXRPTKTLVHPAR-UHFFFAOYSA-N
SMILES: C1N2CN3CN1CP(C2)C3
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)