Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3,5-Triaza-7-Phosphaadamantane

Catalog No. 695467500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-546-7500MG 500 mg
69-546-72G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-546-7500MG Supplier Sigma Aldrich Supplier No. 695467500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3,5-Triaza-7-Phosphaadamantane

Specifications

CAS 53597-69-6
Melting Point 244°C to 250°C
Molecular Formula C6H12N3P
Linear Formula C6H12N3P
MDL Number MFCD00154905
Quantity 500 mg
Synonym 1,3,5-Triaza-7-phosphatricyclo[3.3.1.13.7]decane; NSC 266642; PTA
Molecular Weight (g/mol) 157.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.