CAS RN 536-20-9
CAS RN 536-20-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich Pyridine-2 4 6-tricarboxylic Acid - H3tdc
Sigma Aldrich Pyridine-2 4 6-tricarboxylic Acid - H3tdc
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.