accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClH3CNNH2CH3

CAS RN 536-46-9

IUPAC Name: dihydrogen N1,N4-dimethylbenzene-1,4-diamine dichloride
Synonyms: dihydrogen N1,N4-dimethylbenzene-1,4-diamine dichloride
Molecular Weight (g/mol): 209.11
Molecular Formula: C8H14Cl2N2
InChi Key: PXJHVKRLFWZUNV-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CNC1=CC=C(NC)C=C1
Percent Purity 
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)