Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%

Catalog No. AAA1034314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1034314 25 g
AAA1034322 100 g
AAA1034336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1034314 Supplier Thermo Scientific Chemicals Supplier No. A1034314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 536-46-9 | C8H14Cl2N2 | 209.11 g/mol

It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.

Solubility
Soluble in water

Notes
Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-46-9
Molecular Formula C8H14Cl2N2
Molecular Weight (g/mol) 209.11
MDL Number MFCD00012991
InChI Key IAEDWDXMFDKWFU-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-phenylenediamine dihydrochloride, 4-amino-n,n-dimethylaniline dihydrochloride, n,n-dimethyl-1,4-phenylenediamine dihydrochloride, 1,4-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,4-amino-n,n-dimethylaniline,dihydrochloride, n,n-dimethyl-1,4-benzenediamine dihydrochloride, n,n-dimethyl-p-phenylenediamine 2hcl, dimethyl-p-phenylenediamine dihydrochloride, n,n-dimethyl-p-phenylenediammonium dichloride, pubchem21651
PubChem CID 2724166
IUPAC Name N1,N1-dimethylbenzene-1,4-diamine dihydrochloride
SMILES Cl.Cl.CN(C)C1=CC=C(N)C=C1

Specifications

Melting Point 212°C (decomposition)
Quantity 25 g
UN Number UN2811
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water
Formula Weight 209.12
Percent Purity 98%
Chemical Name or Material N,N-Dimethyl-p-phenylenediamine dihydrochloride
Hazard Category H300+H330-H311-H319
Hazard Statement GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 208-635-7
RTECSNumber ST1300000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.