accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 536-47-0

IUPAC Name: N1,N1-dimethylbenzene-1,4-diamine; sulfuric acid
Synonyms: dimethyl-p-phenylenediamine; sulfuric acid
Molecular Weight (g/mol): 234.27
Molecular Formula: C8H14N2O4S
InChi Key: GLUKPDKNLKRLHX-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.CN(C)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
  • (5)
  • (2)
Quantity 
  • (3)
  • (4)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)