Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethyl-p-phenylenediamine sulfate salt

Catalog No. 18638425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-638-425G 25 g
18-638-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-638-425G Supplier Sigma Aldrich Supplier No. 18638425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N-Dimethyl-p-phenylenediamine sulfate salt

Specifications

CAS 536-47-0
Melting Point 200°C to 205°C (dec.) (lit.)
Boiling Point 495°C (lit.)
Molecular Formula C8H12N2 -+ H2O4S
Linear Formula (CH3)2 NC6H4NH2-+H2SO4
MDL Number MFCD00012992
Quantity 25 g
Synonym 4-(Dimethylamino)aniline sulfate salt; 4-Amino-N,N-dimethylaniline sulfate salt; DMPPDA
Molecular Weight (g/mol) 234.27
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.