accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53608-95-0

IUPAC Name: 2-methoxy-4-methyl-3,4-dihydro-2H-pyran
Synonyms: 2-methoxy-4-methyl-3,4-dihydro-2H-pyran
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: JVTCUFUKBFIIBA-UHFFFAOYNA-N
SMILES: COC1CC(C)C=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results