accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClNOO

CAS RN 5367-28-2

IUPAC Name: 4-chloro-2-methyl-1-nitrobenzene
Synonyms: 4-chloro-2-methyl-1-nitrobenzene
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: NSMZCUAVEOTJDS-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results