Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-nitrotoluene 97.0+%, TCI America™
SDP

Catalog No. C25865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25865G 5 g
C258625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25865G Supplier TCI America Supplier No. C25865G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5367-28-2
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00024579
InChI Key NSMZCUAVEOTJDS-UHFFFAOYSA-N
Synonym 5-chloro-2-nitrotoluene, toluene, 5-chloro-2-nitro, 3-chloro-6-nitrotoluene, benzene, 4-chloro-2-methyl-1-nitro, 2-nitro-5-chlorotoluene, benzene, 1-chloro-3-methyl-4-nitro-9ci, 4-chloro-2-methyl-1-nitro-benzene, acmc-1am5r, 5-chloro-2-nitro-toluene, ksc494a7d
PubChem CID 79329
IUPAC Name 4-chloro-2-methyl-1-nitrobenzene
SMILES CC1=C(C=CC(=C1)Cl)[N+](=O)[O-]

Specifications

Color White-Yellow
Boiling Point 72°C
Quantity 5 g
UN Number 3457
Formula Weight 171.58
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Chloro-2-nitrotoluene
EINECSNumber (3)-0449
RTECSNumber XS9134000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.