accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 537033-55-9

IUPAC Name: 2-(4-chloro-2,6-difluorophenyl)acetic acid
Synonyms: (4-chloro-2,6-difluorophenyl)acetic acid
Molecular Weight (g/mol): 206.57
Molecular Formula: C8H5ClF2O2
InChi Key: LITZKYPXFPYSOE-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results