accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53714-39-9

IUPAC Name: 2-{2,6-dibromo-4-[3,5-dibromo-4-(2-hydroxyethoxy)benzenesulfonyl]phenoxy}ethan-1-ol
Synonyms: 2-{2,6-dibromo-4-[3,5-dibromo-4-(2-hydroxyethoxy)benzenesulfonyl]phenoxy}ethanol
Molecular Weight (g/mol): 653.96
Molecular Formula: C16H14Br4O6S
InChi Key: OFBQIWFJRDGFEK-UHFFFAOYSA-N
SMILES: OCCOC1=C(Br)C=C(C=C1Br)S(=O)(=O)C1=CC(Br)=C(OCCO)C(Br)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results