accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OCaH3COOOH3COOO

CAS RN 53746-45-5

IUPAC Name: calcium bis(2-(3-phenoxyphenyl)propanoate) dihydrate
Synonyms: fenoprofen calcium dihydrate fenoprofen calcium 2H2O fenoprofen calcium salt dihydrate
Molecular Weight (g/mol): 558.64
Molecular Formula: C30H30CaO8
InChi Key: LZPBLUATTGKZBH-UHFFFAOYNA-L
SMILES: O.O.[Ca++].CC(C([O-])=O)C1=CC=CC(OC2=CC=CC=C2)=C1.CC(C([O-])=O)C1=CC=CC(OC2=CC=CC=C2)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Grade 
  • (2)
Industry Type 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)