Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenoprofen calcium salt hydrate, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014520 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4520 5 g
11-101-4519 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4520 Supplier MilliporeSigma Supelco Supplier No. F15175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenoprofen Calcium
Packaging Glass Bottle
CAS 53746-45-5
Physical Form Neat
Linear Formula C30H26O6 Ca
Technique Gas Chromatography
MDL Number MFCD00242722
Synonym (+/-)-2-(3-Phenoxyphenyl)propionic acid
Type Analytical Standard
Molecular Weight (g/mol) 522.60
Industry Type Drug Analysis
Quantity 5 g
Formula Weight 522.60
Percent Purity ≥97% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.