accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53760-27-3

IUPAC Name: N1-(4-aminophenyl)benzene-1,4-diamine; sulfuric acid
Synonyms: N1-(4-aminophenyl)benzene-1,4-diamine; sulfuric acid
Molecular Weight (g/mol): 297.33
Molecular Formula: C12H15N3O4S
InChi Key: OOZQLPDAELLDNY-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=CC=C(NC2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)