Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Diaminodiphenylamine Sulfate (Tech.), 85%, Thermo Scientific™

Catalog No. AC112165000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC112165000 500 g
AC112161000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112165000 Supplier Thermo Scientific Chemicals Supplier No. 112165000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53760-27-3
Molecular Formula C12H15N3O4S
Molecular Weight (g/mol) 297.329
InChI Key OOZQLPDAELLDNY-UHFFFAOYSA-N
Synonym 4,4'-diaminodiphenylamine sulfate, n1-4-aminophenyl benzene-1,4-diamine sulfate, unii-02x5ljd26t, n4-4-aminophenyl benzene-1,4-diamine; sulfuric acid, n1-4-aminophenyl benzene-1,4-diamine; sulfuric acid, 4,4'-iminodianiline sulfate, n-4-aminophenyl benzene-1,4-diamine sulfate 1:1, 4,4'-diaminodiphenylaminsulfat, bis 4-aminophenyl amine sulfate, n-4-aminophenyl benzene-1,4-diamine sulphate 1:1
PubChem CID 104599
IUPAC Name 4-N-(4-aminophenyl)benzene-1,4-diamine;sulfuric acid
SMILES C1=CC(=CC=C1N)NC2=CC=C(C=C2)N.OS(=O)(=O)O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Color Blue
Infrared Spectrum Authentic
Linear Formula (H2NC6H4)2NH.H2SO4
Quantity 500 g
Formula Weight 297.32
Percent Purity ≥80%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Diaminodiphenylamine sulfate, tech., 85%
EINECSNumber 258-748-0
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.