accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH2NOHN

CAS RN 53786-28-0

IUPAC Name: 4-(5-chloro-2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidin-1-ium
Synonyms: 4-(5-chloro-2-oxo-3H-1,3-benzodiazol-1-yl)piperidin-1-ium
Molecular Weight (g/mol): 252.72
Molecular Formula: C12H15ClN3O
InChi Key: DOAYWDKFDPSTSV-UHFFFAOYSA-O
SMILES: ClC1=CC=C2N(C3CC[NH2+]CC3)C(=O)NC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)