Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone 98.0+%, TCI America™
SDP

Catalog No. C30685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30685G 5 g
C306825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30685G Supplier TCI America Supplier No. C30685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53786-28-0
Molecular Formula C12H14ClN3O
Molecular Weight (g/mol) 251.714
MDL Number MFCD02093793
InChI Key DOAYWDKFDPSTSV-UHFFFAOYSA-N
Synonym 5-chloro-1-4-piperidyl-2-benzimidazolinone, 5-chloro-1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 1-4-piperidinyl-1,3-dihydro-5-chlorobenzimidazolone, 5-chloro-1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one, domperidone impurity a, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-benzoimidazol-2-one, 4-5-chloro-2-oxo-2h-benzimidazol-1-yl piperidine, 5-chloro-1-4-piperidyl-3-hydrobenzimidazol-2-one
PubChem CID 104607
IUPAC Name 6-chloro-3-piperidin-4-yl-1H-benzimidazol-2-one
SMILES C1CNCCC1N2C3=C(C=C(C=C3)Cl)NC2=O

Specifications

Melting Point 229°C
Color White
Quantity 5 g
Formula Weight 251.71
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.