accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHN

CAS RN 5379-88-4

IUPAC Name: 3-[(morpholin-4-yl)methyl]-1H-indole
Synonyms: 3-(morpholin-4-ylmethyl)-1H-indole
Molecular Weight (g/mol): 216.28
Molecular Formula: C13H16N2O
InChi Key: VHGISLOQNDQFIW-UHFFFAOYSA-N
SMILES: C(N1CCOCC1)C1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results