Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Morpholinylmethyl)indole, 95%, Thermo Scientific™

Catalog No. AAH6336603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6336603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6336603 Supplier Thermo Scientific Chemicals Supplier No. H6336603

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5379-88-4
Molecular Formula C13H16N2O
Molecular Weight (g/mol) 216.28
MDL Number MFCD00458413
InChI Key VHGISLOQNDQFIW-UHFFFAOYSA-N
Synonym 3-morpholinomethyl indole, indole, 3-morpholinomethyl, 3-morpholin-4-ylmethyl-1h-indole, 4-27-00-00551 beilstein handbook reference, 4-indol-3-ylmethyl morpholine, 3-morpholinomethyl-1h-indole, morpholine, n-3-indolyl methyl, 4-1h-indol-3-ylmethyl morpholine, 3-4-morpholinylmethyl indole
PubChem CID 21477
IUPAC Name 4-(1H-indol-3-ylmethyl)morpholine
SMILES C(N1CCOCC1)C1=CNC2=CC=CC=C12

Specifications

Color White
Quantity 1 g
Beilstein 193005
Formula Weight 216.28
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(4-Morpholinylmethyl)indole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber NM1140000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.