accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOO(E)

CAS RN 538-42-1

IUPAC Name: sodium (2E)-3-phenylprop-2-enoate
Synonyms: sodium cinnamate
Molecular Weight (g/mol): 170.14
Molecular Formula: C9H7NaO2
InChi Key: DXIHILNWDOYYCH-UHDJGPCESA-M
SMILES: [Na+].[O-]C(=O)\C=C\C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results