Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Cinnamate, MP Biomedicals

Catalog No. ICN218375 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218375 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218375 Supplier MP Biomedicals, Inc Supplier No. 0521837583

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 538-42-1
Molecular Formula C9H7NaO2
Molecular Weight (g/mol) 170.14
MDL Number MFCD00064994
InChI Key DXIHILNWDOYYCH-UHDJGPCESA-M
Synonym sodium cinnamate, fortificar, cinnamic acid, sodium salt, unii-1dkd3z3e4y, sodium 3-phenyl-2-propenoate, 2-propenoic acid, 3-phenyl-, sodium salt, 1dkd3z3e4y, cinnamylic acid sodium salt, sodium e-3-phenylprop-2-enoate, potassium cinnamate
PubChem CID 5462638
IUPAC Name sodium (2E)-3-phenylprop-2-enoate
SMILES [Na+].[O-]C(=O)\C=C\C1=CC=CC=C1

Specifications

Quantity 100 g
Formula Weight 170.1
Chemical Name or Material Sodium Cinnamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.