accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH3CNCH3H3CO

CAS RN 538-71-6

IUPAC Name: dodecyldimethyl(2-phenoxyethyl)azanium bromide
Synonyms: domiphen bromide 度米芬
Molecular Weight (g/mol): 414.47
Molecular Formula: C22H40BrNO
InChi Key: OJIYIVCMRYCWSE-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)