Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Domiphen Bromide, BP, 97-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602321 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-321 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-602-321 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D216225KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

D2162, 538-71-6, C22H40BrNO

Spectrum™ Chemical Domiphen Bromide, BP is used as an oral antiseptic. All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 538-71-6
Molecular Formula C22H40BrNO
Molecular Weight (g/mol) 414.47
InChI Key OJIYIVCMRYCWSE-UHFFFAOYSA-M
IUPAC Name dodecyldimethyl(2-phenoxyethyl)azanium bromide
SMILES [Br-].CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1

Specifications

CAS Max % 100%
Loss on Drying 1%
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 97 to 100.5%
Grade BP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Domiphen Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.