CAS RN 539-93-5
IUPAC Name:
3-(dodecanoyloxy)-2-hydroxypropyl dodecanoate
Synonyms:
3-(dodecanoyloxy)-2-hydroxypropyl dodecanoate
Molecular Weight (g/mol):
456.71
Molecular Formula:
C27H52O5
InChi Key:
KUVAEMGNHJQSMH-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCC
1
–
1
of
1
results
1
alpha,alpha'-Dilaurin 96.0+%, TCI America™
CAS: 539-93-5 Molecular Formula: C27H52O5 Molecular Weight (g/mol): 456.708 MDL Number: MFCD00048423 InChI Key: KUVAEMGNHJQSMH-UHFFFAOYSA-N Synonym: Glycerol alpha,alpha′C-Dilaurate PubChem CID: 101758 IUPAC Name: (3-dodecanoyloxy-2-hydroxypropyl) dodecanoate SMILES: CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)O